missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VABP Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
198.00 € - 468.00 €
Specifications
| Antigen | VABP |
|---|---|
| Dilution | Western Blot 1:1000-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
VABP Polyclonal antibody specifically detects VABP in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| VABP | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cancer, Endocrinology, Signal Transduction | |
| PBS (pH 7.3), 50% glycerol | |
| 30820 | |
| IgG | |
| Affinity purified |
| Western Blot 1:1000-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| A-type potassium channel modulatory protein 1, KChIP1, KCHIP1MGC95, Kv channel interacting protein 1, Kv channel-interacting protein 1, potassium channel interacting protein 1, Potassium channel-interacting protein 1, VABP, Vesicle APC-binding protein | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-216 of human KCNIP1 (NP_055407.1). MGAVMGTFSSLQTKQRRPSKDKIEDELEMTMVCHRPEGLEQLEAQTNFTKRELQVLYRGFKNECPSGVVNEDTFKQIYAQFFPHGDASTYAHYLFNAFDTTQTGSVKFEDFVTALSILLRGTVHEKLRWTFNLYDINKDGYINKEEMMDIVKAIYDMMGKYTYPVLKEDTPRQHVDVFFQKMDKNKDGIVTLDEFLESCQEDDNIMRSLQLFQNVM | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title