missing translation for 'onlineSavingsMsg'
Learn More

Vaccenic Acid Methyl Ester, TRC

Product Code. 30137495
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30137495 1 g 1g
30003725 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30137495 Supplier Toronto Research Chemicals Supplier No. TRCV0700101G
 Restricted Item
Estimated Shipment: 01-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6198-58-9
Molecular Formula C19 H36 O2
Molecular Weight (g/mol) 296.49
Synonym trans-11-Octadecanoic acid methyl ester
IUPAC Name methyl (E)-octadec-11-enoate
SMILES CCCCCC\C=C\CCCCCCCCCC(=O)OC

Specifications

Quantity 1 g
InChI Formula InChI=1S/C19H36O2/c1-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19(20)21-2/h8-9H,3-7,10-18H2,1-2H3/b9-8+
Formula Weight 296.2715
Chemical Name or Material Methyl-trans-vaccenate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.