missing translation for 'onlineSavingsMsg'
Learn More

Cytiva VacuCap™ 90 Vacuum Filtration Devices with Individually Attached Tubing

Product Code. 17124821
Change view
Click to view available options
Pore Size:
0.2 μm
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Pore Size Unit Size
17124821 0.2 μm Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17124821 Supplier Cytiva Supplier No. TA4622
  • ONLINE EXCLUSIVE PRICE
    324.00 €
    301.87 € / Pack of 10
    Save 22.13 €
    6% Off
Estimated Shipment: 05-03-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Remove endotoxin from water, buffer, neutral sugars, and certain biological solutions.

  • Not approved for use in medical, clinical, surgical or other patient applications
TRUSTED_SUSTAINABILITY

Specifications

Material (Membrane) Hydrophilic PES
Pore Size 0.2 μm
Sterility Sterile
Filtration Area 60 cm2
No. per Pack 10/Pk.
Diameter (Metric) 90 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.