missing translation for 'onlineSavingsMsg'
Learn More

Greiner Bio-One VacuDrop™ Blood Tube Piercer

Product Code. 13058782
Change view
Click to view available options
Unit Size:
Pack of 300
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Unit Size
13058782 Pack of 300
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13058782 Supplier Greiner Bio-One Supplier No. 458815

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The Greiner Bio-One™ VacuDrop™ Blood Tube Piecer is used to collect smears of blood without having to open Vacuette™ blood collection tubes for even safer and hygienic handling.

  • Dedicated plastic spikes used in combination with Vacuette™ blood collection tubes
  • Permits collection of smears of blood without having to open tubes
  • Ensures safe and hygienic handling of blood samples with reduced risk of infection
  • Specially designed for haematological labs for differential blood counts
TRUSTED_SUSTAINABILITY

Specifications

Autoclavable Not Autoclavable
Color Clear
Disposable Single-use
For Use With (Equipment) Vacuette™ Blood Tubes
Material Plastic
Product Type VacuDrop Spike
Sterility Non-sterile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.