missing translation for 'onlineSavingsMsg'
Learn More

Greiner Bio-One Vacuette™ Low Volume Blood Collection Tubes

Product Code. 15393831
Change view
Click to view available options
Closure Color:
Lavender
Unit Size:
Pack of 1200
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Closure Color Unit Size
15393831 Lavender Pack of 1200
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15393831 Supplier Greiner Bio-One Supplier No. 454052
  • ONLINE EXCLUSIVE PRICE
    261.00 €
    213.03 € / Pack of 1200
    Save 47.97 €
    18% Off
 Restricted Item
Estimated Shipment: 21-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Product Type Low Volume Blood Collection Tube
Additive K2 EDTA
Material PET
Sterility Sterile
Capacity (Metric) 1 mL
Closure Color Lavender
Tube Size 13 x 75 mm
For Use With (Application) Blood Collection
compliance-icons
Product Identifier
  • VACUETTE K2E K2EDTA Sep Blood Collection Tube
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: Ethylendiaminetetraacetic Acid Dipotassium Dihydrate K2EDTA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.