missing translation for 'onlineSavingsMsg'
Learn More

BD Vacutainer™ Citrate Tube

Product Code. 12997656
Change view
Click to view available options
Capacity (Metric):
1.8 mL
2.7 mL
4.5 mL
Material:
Glass
PET
Unit Size:
Pack of 100
Pack of 1000
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Material unitSize
12997656 2.7 mL PET Pack of 1000
16425264 2.7 mL PET Pack of 100
12967656 4.5 mL Glass Pack of 1000
12987656 1.8 mL PET Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 12997656 Supplier BD Supplier No. 363095
 Restricted Item
Estimated Shipment: 28-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Contains buffered trisodium citrate solution. BD™ Vacutainer™ Citrate Tube contains an anticoagulant additive equivalent to 3.2% trisodium citrate for coagulation investigations. It is also suitable for carrying out special test procedures, such as the platelet function test PFA-100.

Trisodium citrate works as an anticoagulant by forming complexes with metal ions such as calcium, thereby inhibiting the coagulation cascade. Anticoagulation with trisodium citrate is reversible. The blood to additive ratio is 9:1.

BD Vacutainer™ Citrate Tubes are available in glass or Plus PET, both with blue Hemogard™ closures.

  • Both tubes feature clinically equivalent performance 
  • Glass tubes are recognized as the global “Gold Standard” and have an internal coating of a special silicone to minimize contact activation
  • Plus PET tubes include a Minimum Fill Indicator marking and feature innovative tube geometry that minimizes tube headspace and associated platelet activation (to optimize APTT monitoring of unfractionated heparin patients)
  • Glass tubes are not recommended for centrifugation at more than 2200 g in a swing-out rotor (1300 g for a fixed angle rotor), and so may not be appropriate for use with platelet free plasma
TRUSTED_SUSTAINABILITY

Specifications

Product Type Venous Blood Sampling Coagulation Tube
Additive Sodium Citrate
Material PET
Closure Type Hemogard
Capacity (Metric) 2.7 mL
Closure Color Light Blue
Height (Metric) 75 mm
For Use With (Application) Used for coagulation studies
Label Type Block
Diameter (Metric) Outer 13 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.