Learn More
BD Vacutainer™ Eclipse™ PST™ II Plasma Separator Tubes
Description
During centrifugation of Eclipse PST II tubes, an inert gel separates serum and red blood cells, preventing the contamination of plasma from the separated cellular components. Routine analytes (e.g., potassium, glucose) and special chemistries (e.g., therapeutic drugs, proteins, peptides, steroids and vitamins) demonstrate a high degree of stability.
- The main advantages of gel tubes versus non-gel tubes are:
- Stable barrier between plasma and red blood cells
- Better analyte stability and sample quality
- Optimized workflow: short centrifugation time, sample processing and archiving in one tube
- No possibility of confusion due to the use of secondary tubes
- Storage: Store at 4-25°C, protected from direct sunlight; optimum separation is achieved at 20-25°C
- Mixing the tube: Invert tube 8-10 times immediately after collecting sample to avoid microclotting
- Centrifugation conditions at 18-25°C: 1300-2000g for 10 minutes or 3000g for 5 minutes
Specifications
Specifications
| Product Type | PST II Tube |
| Additive | Lithium Heparin and Plasma Separator |
| Material | PET |
| Sterility | Sterile |
| Closure Type | Hemogard |
| Capacity (Metric) | 4.5 mL |
| For Use With (Equipment) | Vacutainer™ Needle and Tube Holder; Centrifugator |
| Closure Color | Light Green |
| Tube Size | 13 x 100 mm |
| For Use With (Application) | Used to obtain and separate a serum sample |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.