missing translation for 'onlineSavingsMsg'
Learn More

BD Vacutainer™ Glass ACD Solution Tube with Yellow Closure

Product Code. 12937696
Change view
Click to view available options
Capacity (Metric):
6 mL
8.5 mL
Additive:
ACD Solution A
ACD Solution B
Closure Type:
Hemogard
Traditional Cap
Unit Size:
Pack of 100
Pack of 1000
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Additive Closure Type unitSize
12937696 8.5 mL ACD Solution A Traditional Cap Pack of 1000
15178988 8.5 mL ACD Solution A Traditional Cap Pack of 100
12947696 6 mL ACD Solution B Hemogard Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 12937696 Supplier BD Supplier No. 366645
 Restricted Item
Estimated Shipment: 03-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Used to obtain a whole blood or a plasma sample. BD Vacutainer™ Glass ACD Solution Tube with Yellow Closure contains the active anticoagulant Tri-sodium Citrate with Citric Acid and Dextrose. Two formulations of the ACD solution are available.

BD Vacutainer™ ACD tubes are available in glass only and are identifiable by a yellow cap (Hemogard™ or conventional). They contain the active anticoagulant Tri-sodium Citrate with Citric Acid and Dextrose in one of two formulations. The concentration of additives in each solution is as follows:
  • ACD Solution A (conventional closure)
    • Sodium citrate: 22.0g/L
    • Dextrose: 24.5g/L
    • Citric acid: 8.0g/L
    • Antimycotic (K sorbate) reagent: 0.15g/L
  • ACD Solution B (Hemogard™ closure)
    • Sodium citrate: 13.2g/L
    • Dextrose: 14.7g/L
    • Citric acid: 4.8g/L
    • Antimycotic (K sorbate) reagent: 0.2g/L
TRUSTED_SUSTAINABILITY

Specifications

Product Type ACD Solution A Tube
Additive ACD Solution A
Material Glass
Sterility Sterile
Closure Type Traditional Cap
Capacity (Metric) 8.5 mL
Closure Color Yellow
Tube Size 16 x 100 mm
For Use With (Application) Used to obtain a whole blood or plasma sample
Label Type Paper
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.