missing translation for 'onlineSavingsMsg'
Learn More

BD Vacutainer™ Eclipse™ Blood Collection Needle

Product Code. 12399299
Change view
Click to view available options
Gauge:
21 G
22 G
Color:
Black
Green
Unit Size:
Pack of 480
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Gauge Color unitSize
12399299 21 G Green Pack of 480
12977716 22 G Black Pack of 480
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 12399299 Supplier BD Supplier No. 368609
 Restricted Item
Estimated Shipment: 12-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Built-in safety shield is aligned with the bevel of the needle. BD Vacutainer™ Eclipse™ Blood Collection Needle has a safety shield that permanently locks into place, significantly reducing the risk of needle stick injuries.

The BD Vacutainer™ system is a closed system optimally designed and tested for controlling pre-analytical variability throughout the specimen collection and handling process.

The BD Vacutainer™ Eclipse™ Blood Collection Needle for venous blood sampling has a fully integrated safety shield over the needle, which once activated protects against needle stick injuries.

  • Safety shield is an integral part of the needle and is aligned to the needle bevel
  • Safety shield engages over the needle with an audible click, irreversibly locking with a triple closure mechanism
  • The safety mechanism is designed for single handed activation
  • Color coded shield corresponds to needle gauge size
  • Designed for use with Vacutainer™ tube holders and evacuated tubes
TRUSTED_SUSTAINABILITY

Specifications

Product Type Blood Collection Needle
Color Green
For Use With (Application) Venous Blood Sampling
For Use With (Equipment) Vacutainer™ blood collection tubes and tube holders
Gauge 21 G
Includes Integrated Safety Shield
Length (English) 1.25 in.
Length (Metric) 32 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.