missing translation for 'onlineSavingsMsg'
Learn More

BD Vacutainer™ Microbiology Tube C & S

Product Code. 12927776
Change view
Click to view available options
Volume (Metric):
10 mL
Unit Size:
Pack of 1000
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
12927776 10 mL Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12927776 Supplier BD Supplier No. 364955
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
round bottom with BD Hemogard™ closure plastic for the collection & transport of urine specimens green volume 10mL 16mm x 100m BD Vacutainer
TRUSTED_SUSTAINABILITY

Specifications

Bottom Shape Round
Diameter (Metric) Inner 16 mm
For Use With (Application) Collection and transport of urine specimens
Height (Metric) 100 mm
Volume (Metric) 10 mL
compliance-icons
Product Identifier
  • TUBE URIN PLH 16X100 10 C&S KAGN CE
Signal Word
  • Danger
Hazard Category
  • Reproductive toxicity Category 1B
Hazard Statement
  • H360FD-May damage fertility. May damage the unborn child.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Borax
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.