missing translation for 'onlineSavingsMsg'
Learn More

BD Vacutainer™ SST™ II Advance Tubes

Product Code. 12927696
Change view
Click to view available options
Capacity (Metric):
3.5 mL
5 mL
8.5 mL
Closure Color:
Gold
Red
Label Type:
Block
Paper
Unit Size:
Pack of 1000
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Closure Color Label Type Unit Size
12927696 8.5 mL Gold Block Pack of 1000
12917696 5 mL Gold Block Pack of 1000
12987686 3.5 mL Gold Block Pack of 1000
12997686 3.5 mL Red Block Pack of 1000
12907696 5 mL Red Paper Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 12927696 Supplier BD Supplier No. 367958
Save Up to 60% on Microbiology Essentials
Use promo code 28004
View all promotions
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Prevents the contamination of the serum from the separated cellular components. BD Vacutainer™ SST™ II Advance Tubes feature an inert, stable gel that separates the serum and the blood clot during centrifugation. The type of gel used enables short centrifugation times.

Using BD SST™ II Advance, routine analytes in clinical chemistry, such as potassium and glucose, are still stable after a week of storage at 2–8°C. Clinical evaluation of special chemistries (such as therapeutic drugs, proteins, peptides, steroids and vitamins) demonstrates a high degree of stability with the acrylic gel in these tubes, which is a distinct advantage during transport and storage.

Advantages of gel tubes versus non-gel tubes:

  • Stable barrier between serum and clotted blood, therefore better analyte stability and sample quality
  • Optimization of the work flow: short centrifugation time, sample processing and archiving in the primary tube
  • No possibility of misidentification due to the use of secondary tubes

Tube usage guidelines:

  • Clotting Times: the minimum recommended coagulation time for patients who have not received anti-coagulation treatment is 30 minutes. Tubes contain a clot activator of silica particles.
  • Centrifugation Conditions: 1300–2000 g for 10 minutes or alternately 3000 g for 5 minutes, at 18–25°C.
  • Effects of Temperature: cooling of the tube during centrifuging can affect the movement capability of the gel. Optimum separation of serum and coagulated blood is achieved at 20–25°C.
  • Store at 4–25°C and protected from direct sunlight during storage.
TRUSTED_SUSTAINABILITY

Specifications

Product Type Serum Separator Tube
Additive Silica
Material PET
Sterility Sterile
Closure Type Hemogard
Capacity (Metric) 8.5 mL
Closure Color Gold
Height (Metric) 100 mm
For Use With (Application) Used to obtain and separate a serum sample
Label Type Block
Diameter (Metric) Outer 16 mm
Certifications/Compliance CE
Show More Show Less
Product Title
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.