Learn More
VACUUBRAND™ Vacuubrand Operating Module C3-B For VACUU·BUS™ Plug Connection
Product Code. 10576855
Click to view available options
For Use With:
VACUU·BUS Plug Connection
Unit Size:
Each
Product Code. 10576855
Brand: VACUUBRAND™ 20636668
Only null left
Add to basket
Add to basket
The upgrade kit SYNCHRO for a second vacuum connection can be equipped, depending on the application, with a manual flow control valve (677137) or an in-line solenoid valve (636668) for electronic vacuum control with the CVC 3000 vacuum controller. These funtional elements for individual vacuum control are necessary to operate the vacuum ports.
TRUSTED_SUSTAINABILITY
Specifications
For Use With VACUU·BUS Plug Connection
Plug Type VACUU·BUS™ Plug
Quantity 1 Each
Type Operating Module C3-B