Learn More
VACUUBRAND™ VACUUBUS™ CVC3000 Vacuum Controller Solenoid Valves
Description
- State-of-the-art vacuum gauges for laboratory and industrial operation have to meet high standards in terms of gauge head resistance to chemicals, reliability in rugged applications and operator-friendly interfaces. VACUUBRAND has a comprehensive range of electronic measuring instruments for the range of atmospheric pressure to 5 x 10-9 mbar.
- by cyclically turning the pump on/off (with CVC 3000 controller with module VMS-B)
- by cyclically opening/closing an in-line solenoid valve (CVC 3000) or
- by changing the pump ́s motor speed continuously (VARIO™ pumps in connection with CVC 3000)
Specifications
Specifications
| For Use With | CVC3000 vacuum controller and DCP3000 vacuum gauge |
| Type | Solenoid Isolation Valve |