Learn More
Memmert™ Vacuum Oven Thermoshelf
Product Code. 11901459 Shop All Memmert Products
Change view
Click to view available options
For Use With (Equipment):
Model 200
Model 400
Model 500
Material:
Aluminum
Stainless Steel
Unit Size:
Each
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Material Unit Size
11901459 Model 400 Stainless Steel Each
11741464 Model 200 Aluminum Each
11751464 Model 400 Aluminum Each
11991449 Model 200 Stainless Steel Each
11323692 Model 500 Aluminum Each
11911459 Model 500 Stainless Steel Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 11901459 Supplier Memmert™ Supplier No. VO49B00734
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Specifications
Certifications/Compliance Calibration certificate
For Use With (Equipment) Model 400
Type Shelf
For Use With (Application) Individual overtemperature protection for each shelf MLOP (Multi-Level-Overtemperature-Control)
Includes Local Temperature Sensing (Pt100, 4-Wire-Circuit)
Material Stainless Steel