missing translation for 'onlineSavingsMsg'
Learn More

Vadadustat, MedChemExpress

Product Code. p-200098019
Encompass_Preferred
Click to view available options
Quantity:
2 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
2mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16483318

Brand: Medchem Express HY101277

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Vadadustat (PG-1016548) is a titratable, oral hypoxia-inducible factor prolyl hydroxylase (HIF-PH) inhibitor. Vadadustat is an erythropoiesis-stimulating agent and has the potential for anemia treatment in chronic kidney disease in vivo.

Vadadustat (PG-1016548) is a titratable, oral hypoxia-inducible factor prolyl hydroxylase (HIF-PH) inhibitor. Vadadustat is an erythropoiesis-stimulating agent and has the potential for anemia treatment in chronic kidney disease in vivo.

Specifications

Chemical Name or Material Vadadustat
CAS 1000025-07-9
Color Off-White
Purity Grade Notes Research
Molecular Formula C14H11ClN2O4
Quantity 10 mg
Synonym PG-1016548 AKB-6548
Solubility Information DMSO : 100 mg/mL (326.05 mM; Need ultrasonic)
SMILES O=C(O)CNC(C1=NC=C(C2=CC=CC(Cl)=C2)C=C1O)=O
Molecular Weight (g/mol) 306.7
Formula Weight 306.7
Percent Purity 99.25%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.