Learn More
Julabo Valegro Ethernet Activation
Product Code. 30244965
Change view
Click to view available options
Connection Type:
Ethernet
Unit Size:
Each
Product Code. Connection Type Unit Size
30244965 Ethernet Each
1 options
Product Code. 30244965 Supplier Julabo Supplier No. 30004147
 Restricted Item
Estimated Shipment: 14-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Activation of the Valegro Ethernet to allow integration into the network.
Activating the Ethernet interface makes it possible to integrate Valegro recirculating coolers into your network and thus gain full access to all their functions via a PC.
The Ethernet interface already installed in the Valegro series can be activated at a later date.
Specifications
Type Ethernet Activation
Connection Type Ethernet
Product Line Valegro