missing translation for 'onlineSavingsMsg'
Learn More

Emma Safety Footwear Valentino S3 Shoe

Product Code. 17592446
Change view
Click to view available options
Size:
39
40
41
42
43
44
45
46
47
48
Unit Size:
1 pair
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
17592446 41 1 pair
17572446 39 1 pair
17582446 40 1 pair
17502456 42 1 pair
17512456 43 1 pair
17522456 44 1 pair
17532456 45 1 pair
17542456 46 1 pair
17552456 47 1 pair
17562456 48 1 pair
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
This item is not returnable. View return policy
Product Code. 17592446 Supplier Emma Safety Footwear Supplier No. MM16400041
  • ONLINE EXCLUSIVE PRICE
    237.07 €
    170.21 € / 1 pairs
    Save 66.86 €
    28% Off
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Break new ground in the Valentino Frontier, the executive safety shoe that literally has no boundaries.

  • Industries:
    • Chemical industry
    • Government & services
    • Oil & gas
  • Safety shoe
    • S3-Safetyshoe with sole plate for wet work

Specifications

Certifications/Compliance 20345:2011
Closure Type Laces
Color Black
For Use With (Application) Chemical Industry, Government and Services, Oil and Gas
Material Leather
Size 41
Toe Material Steel
Fit Extra-wide Width
Ankle Style High
Product Type Safety Shoe
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.