missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Valeric Acid, 99%

Product Code. 10686584 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 mL
100 mL
1 L
2.5 L
Packaging:
Glass bottle
Unit Size:
1 L
100mL
2.5 L
5mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10686584 100 mL Glass bottle 100mL
10123100 5 mL Glass bottle 5mL
10316600 1 L Glass bottle 1 L
10686394 2.5 L Glass bottle 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10686584 Supplier Thermo Scientific Chemicals Supplier No. 149571000
Spotlight Offers
Use promo code 28038
View all promotions
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 109-52-4 | C5H10O2 | 102.133 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109-52-4
Molecular Formula C5H10O2
Molecular Weight (g/mol) 102.133
MDL Number MFCD00004413
InChI Key NQPDZGIKBAWPEJ-UHFFFAOYSA-N
Synonym valeric acid, n-valeric acid, n-pentanoic acid, valerianic acid, 1-butanecarboxylic acid, propylacetic acid, butanecarboxylic acid, pentoic acid, kyselina valerova, valeric acid, n
PubChem CID 7991
ChEBI CHEBI:17418
IUPAC Name pentanoic acid
SMILES CCCCC(=O)O

Specifications

Melting Point -20.0°C to +18.0°C
Boiling Point 185.0°C
Color Colorless to Yellow
Density 0.9400g/mL
Quantity 100 mL
Flash Point 86°C
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Liquid
Refractive Index 1.4070 to 1.4100
Assay Percent Range 99%
Specific Gravity 0.94
Linear Formula CH3(CH2)3CO2H
Beilstein 02, 299
Merck Index 15, 10090
Solubility Information Solubility in water: 40g/L (20°C). Other solubilities: soluble in alcohol and ether
Formula Weight 102.13
Percent Purity 99%
Chemical Name or Material Valeric acid
Show More Show Less
compliance-icons
Product Identifier
  • Valeric acid
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.