missing translation for 'onlineSavingsMsg'
Learn More

gamma-Valerolactone, 98%

Product Code. 10026003 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 mL
100 mL
500 mL
Packaging:
Glass bottle
Unit Size:
100mL
500mL
5mL
This item is not returnable. View return policy

Product Code. 10026003

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 108-29-2 | C5H8O2 | 100.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-29-2
Molecular Formula C5H8O2
Molecular Weight (g/mol) 100.12
MDL Number MFCD00005400
InChI Key GAEKPEKOJKCEMS-UHFFFAOYSA-N
Synonym gamma-valerolactone, 4-valerolactone, 4-pentanolide, 5-methyldihydrofuran-2 3h-one, 4-hydroxypentanoic acid lactone, gamma-pentalactone, 4-hydroxyvaleric acid lactone, 2 3h-furanone, dihydro-5-methyl, 4-methyl-gamma-butyrolactone, pentanolide-1,4
PubChem CID 7921
ChEBI CHEBI:48569
IUPAC Name 5-methyloxolan-2-one
SMILES CC1CCC(=O)O1

Specifications

Melting Point -31.0°C
Color Colorless
Density 1.0500g/mL
Boiling Point 207.0°C to 208.0°C
Flash Point 96°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4320 to 1.4340
Quantity 500 mL
Beilstein 17, 235
Specific Gravity 1.05
Solubility Information Solubility in water: miscible. Other solubilities: miscible with most common organic solvents,resins,propylene glycol,oil
Viscosity 2.18 mPa.s (25°C)
Formula Weight 100.12
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material γ-Valerolactone
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.