missing translation for 'onlineSavingsMsg'
Learn More

Valerophenone, 98%

Product Code. 15149183 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15149183 500 g Glass bottle 500g
10212752 5 g Glass bottle 5g
10497150 25 g Glass bottle 25g
10626152 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 15149183 Supplier Thermo Scientific Acros Supplier No. 140805000
 Restricted Item
Estimated Shipment: 11-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1009-14-9 | C11H14O | 162.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1009-14-9
Molecular Formula C11H14O
Molecular Weight (g/mol) 162.23
MDL Number MFCD00009480
InChI Key XKGLSKVNOSHTAD-UHFFFAOYSA-N
Synonym valerophenone, butyl phenyl ketone, pentanophenone, 1-phenyl-1-pentanone, 1-pentanone, 1-phenyl, unii-f27q043nt1, n-butyl phenyl ketone, 1-benzoylbutane, valerophenone, 8ci
PubChem CID 66093
ChEBI CHEBI:36812
IUPAC Name 1-phenylpentan-1-one
SMILES CCCCC(=O)C1=CC=CC=C1

Specifications

Melting Point -9.0°C
Color Green-Yellow to Yellow
Density 0.9800g/mL
Boiling Point 105.0°C to 107.0°C (5.0 mmHg)
Flash Point 102°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula C6H5CO(CH2)3CH3
Refractive Index 1.5143 to 1.5163
Quantity 500 g
Beilstein 07,327
Specific Gravity 0.98
Formula Weight 162.23
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Valerophenone, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.