missing translation for 'onlineSavingsMsg'
Learn More

Valerophenone, 99.26%, MP Biomedicals™

Product Code. 11459741 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11459741 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11459741 Supplier MP Biomedicals Supplier No. 0215826725
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 1009-14-9
Color Colorless
Density 0.988g/mL
Assay Percent Range 0.9926
Molecular Formula C11H14O
MDL Number MFCD00009480
Quantity 25 g
Synonym valerophenone, butyl phenyl ketone, pentanophenone, 1-phenyl-1-pentanone, 1-pentanone, 1-phenyl, unii-f27q043nt1, n-butyl phenyl ketone, 1-benzoylbutane, valerophenone, 8ci
InChI Key XKGLSKVNOSHTAD-UHFFFAOYSA-N
SMILES CCCCC(=O)C1=CC=CC=C1
IUPAC Name 1-phenylpentan-1-one
Molecular Weight (g/mol) 162.23
PubChem CID 66093
ChEBI CHEBI:36812
Formula Weight 162.23
Physical Form Liquid
Chemical Name or Material Valerophenone, For GC analysis
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.