missing translation for 'onlineSavingsMsg'
Learn More

Valeryl chloride, 98%

Product Code. 10194320 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 g
1 kg
Packaging:
Glass bottle
Unit Size:
100g
1kg
This item is not returnable. View return policy

Product Code. 10194320

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 638-29-9 | C5H9ClO | 120.58 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 638-29-9
Molecular Formula C5H9ClO
Molecular Weight (g/mol) 120.58
MDL Number MFCD00000757
InChI Key XGISHOFUAFNYQF-UHFFFAOYSA-N
Synonym valeryl chloride, valeroyl chloride, valerylchloride, n-valeryl chloride, n-pentanoyl chloride, n-valeroyl chloride, pentanoic acid chloride, n-pentoyl chloride, valerylchlorid, valeroylchloride
PubChem CID 61186
IUPAC Name pentanoyl chloride
SMILES CCCCC(Cl)=O

Specifications

Melting Point -110.0°C
Color Colorless to Yellow
Density 0.9900g/mL
Boiling Point 125.0°C to 127.0°C
Flash Point 32°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3(CH2)3COCl
Refractive Index 1.4190 to 1.4210
Quantity 100 g
Beilstein 02,301
Specific Gravity 0.99
Solubility Information Solubility in water: hydrolysis. Other solubilities: soluble in most common organic solvents
Formula Weight 120.58
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Valeryl chloride, 98%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Valeryl chloride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 B
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Substances and mixtures corrosive to metals Category 1
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H331-Toxic if inhaled.
  • H226-Flammable liquid and vapour.
  • H290-May be corrosive to metals.
  • H412-Harmful to aquatic life with long lasting effects.
  • EUH029-Contact with water liberates toxic gas.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.