Learn More
Thermo Scientific™ NanoDrop™ Cuvette Qualification Filter Kit
Product Code. 31921092 Shop All Thermo Scientific Products
Change view
Click to view available options
For Use With (Equipment):
NanoDrop UltraC, NanoDrop UltraC FL, NanoDrop OneC, and NanoDrop 2000C Spectrophotometers
Unit Size:
Each
Product Code. For Use With (Equipment) Unit Size
31921092 NanoDrop UltraC, NanoDrop UltraC FL, NanoDrop OneC, and NanoDrop 2000C Spectrophotometers Each
1 options
Product Code. 31921092 Supplier Thermo Scientific™ Supplier No. 840257300
 Restricted Item
Estimated Shipment: 11-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
The Cuvette Qualification Filter Kit for the cuvette based Thermo Scientfic™ NanoDrop™ UltraC and NanoDrop™ UltraC FL, NanoDrop™ OneC and NanoDrop™ 2000c Spectrophotometers is an essential tool for ensuring the optimal performance of your spectrophotometers. This kit provides you with the necessary filters for accurate cuvette qualification testing.
Includes: 1 x 10%T Filter (Calibrated), 1 x 50%T (Calibrated), and a certificate with traceability and NVLAP 17025, Lab Code 200955, accreditation.
Specifikationer
Description Qualification Filter Kit for NanoDrop OneC and NanoDrop 2000C Spectrophotometers
For Use With (Equipment) NanoDrop UltraC, NanoDrop UltraC FL, NanoDrop OneC, and NanoDrop 2000C Spectrophotometers
Includes 1 x Filter holder blank, 1 x 10%T Filter, Calibrated, 1 x 50%T Filter, Calibrated, Certificate with traceability and NVLAP 17025, Lab Code 200955, accreditation
Produkttitel
Välj ett problem
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.