missing translation for 'onlineSavingsMsg'
Learn More

Valifenalate, TRC

Product Code. 30137038
Change view
Click to view available options
Quantity:
50 mg
100 mg
250 mg
Unit Size:
100mg
250mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30137038 250 mg 250mg
30138341 50 mg 50mg
17932175 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137038 Supplier Toronto Research Chemicals Supplier No. TRCV094030250MG
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 283159-90-0
Molecular Formula C19 H27 Cl N2 O5
Molecular Weight (g/mol) 398.88
Synonym β-Alanine, N-[(1-methylethoxy)carbonyl]-L-valyl-3-(4-chlorophenyl)-, methyl ester, IR 5885, Valifenalate, Valiphenal
IUPAC Name methyl 3-(4-chlorophenyl)-3-[[(2S)-3-methyl-2-(propan-2-yloxycarbonylamino)butanoyl]amino]propanoate
SMILES COC(=O)CC(NC(=O)[C@@H](NC(=O)OC(C)C)C(C)C)c1ccc(Cl)cc1

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C19H27ClN2O5/c1-11(2)17(22-19(25)27-12(3)4)18(24)21-15(10-16(23)26-5)13-6-8-14(20)9-7-13/h6-9,11-12,15,17H,10H2,1-5H3,(H,21,24)(H,22,25)/t15?,17-/m0/s1
Formula Weight 398.1608
Chemical Name or Material Valifenalate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.