missing translation for 'onlineSavingsMsg'
Learn More

Valine Ethyl Ester, Hydrochloride, TRC

Product Code. 30450913
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30450913 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30450913 Supplier Toronto Research Chemicals Supplier No. TRCV0943101G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Valine Ethyl Ester','Hydrochloride
CAS 23358-42-1
Molecular Formula C7 H15 N O2 . Cl H
Quantity 1 g
Synonym Valine','ethyl ester','hydrochloride (1:1), DL-Valine','ethyl ester','hydrochloride, Valine','ethyl ester','hydrochloride (9CI), Valine','ethyl ester','hydrochloride','DL- (8CI), (±)-Valine ethyl ester hydrochloride, Ethyl DL-valinate hydrochloride, O-Ethyl-DL-valine hydrochloride, DL-Valine ethyl ester hydrochloride, Ethyl 2-amino-3-methylbutanoate hydrochloride, 2-Amino-3-methylbutanoic acid ethyl ester hydrochloride
InChI Formula InChI=1S/C7H15NO2.ClH/c1-4-10-7(9)6(8)5(2)3;/h5-6H,4,8H2,1-3H3;1H
SMILES Cl.CCOC(=O)C(N)C(C)C
IUPAC Name ethyl 2-amino-3-methylbutanoate;hydrochloride
Molecular Weight (g/mol) 181.66
Formula Weight 181.087
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.