missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies Valproic Acid (Sodium Salt)
Description
- VaLproic Acid (VPA) is a short-chain fatty acid that acts as an epigenetic modifier by inhibiting Histone Deacetylases (HDACs) with IC50 values ranging from 0.4 - 3 mM
- VPA can also increase γaminobutyric acid (GABA) levels via inhibition of enzymes involved in GABA metabolism
- Other effects include depletion of cellular inositol by inhibiting myo-inositol-1-phosphate synthase
- (Gottlicher et al.), Khan et al.), Phiel et al.), Rosenberg)
- Enables chemical reprogramming (without genetic factors) of mouse embryonic fibroblasts to induced pluripotent stem (iPS) cells, in combination with CHIR99021, Forskolin, Tranylcypromine, 3-Deazaneplanocin A, and E-616452 (Hou et al.)
- Increases the reprogramming efficiency of mouse embryonic fibroblasts to iPS cells (Huangfu et al. 2008a)
- Promotes reprogramming of human fibroblasts to iPS cells using only 2 factors, OCT4 and SOX2 (Huangfu et al. 2008b)
- Direct lineage reprogramming of fibroblasts to mature neurons, in combination with CHIR99021, RepSox, Forskolin, SP600125, Gö6983 and Y-27632 (Hu et al.)
- Mediates ex vivo expansion of cord blood CD34+ hematopoietic stem and progenitor cells (Chaurasia et al.)
- Promotes the proliferation and self-renewal of human and mouse hematopoietic progenitor cells (Bug et al.), De Felice et al.)
- Promotes differentiation of neurons and suppresses differentiation of astrocytes and oligodendrocytes from rat neural progenitor cells (Hsieh et al.), Jung et al.)
- Promotes osteogenic differentiation of human mesenchymal stem cells (Cho et al.)
Specifications
Specifications
| CAS | 1069-66-5 |
| Synonym | 2-Propylvaleric acid; Sodium valproate; VPA |
| Quantity | 500 mg |
| Molecular Formula | C8H15O2 · Na |
| Purity | ≥95% |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?