missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Valve and Electrical Lockout Kit

Product Code. 12798019 Shop All Brady Products
Change view
Click to view available options
Product Type:
Ultimate Lockout Kit
Valve and Electrical Lockout Kit
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Unit Size
12798019 Valve and Electrical Lockout Kit Each
11756795 Ultimate Lockout Kit Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 12798019 Supplier Brady™ Supplier No. 806174
  • ONLINE EXCLUSIVE PRICE
    1085.00 €
    1027.96 € / Pack of 1
    Save 57.04 €
    5% Off
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This combination Valve and Electrical Lockout Kit gives you everything you need to lock out the most common mechanical and electrical control points.

  • Our combination lockout kit gives you everything you need to lock out the most common mechanical and electrical control points.

Specifications

Product Type Valve and Electrical Lockout Kit
Alternate Part Number VALVE & ELECTRICAL LOCKOUT KIT (EN)
Color Red/Silver
For Use With (Application) Lockout Tagout
Includes Large Universal Valve Lockout Base Clamping Unit
Quantity 1 Ea.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.