Learn More
Brady™ Valve and Electrical Lockout Kit
Product Code. 11756795
Click to view available options
Product Type:
Ultimate Lockout Kit
Valve and Electrical Lockout Kit
missing translation for 'unitSize'
Each
Product Code. 11756795
missing translation for 'mfr': Brady™ 806186
Only null left
Add to basket
Add to basket
This combination Valve and Electrical Lockout Kit gives you everything you need to lock out the most common mechanical and electrical control points.
  • Our combination lockout kit gives you everything you need to lock out the most common mechanical and electrical control points.
Specifications
Product Type Ultimate Lockout Kit
Alternate Part Number Valve and Electrical Lockout Kit (FF)
For Use With (Application) Lockout Tagout
Includes Large Fixing Device, Large Locking Arm, Cable Tie
Quantity 1 Ea.