missing translation for 'onlineSavingsMsg'
Learn More

Vanadium Chloride, TRC

Product Code. 30455007
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
Product Code. Quantity unitSize
30455007 10 g 10g
1 options
This item is not returnable. View return policy

Product Code. 30455007

Brand: Toronto Research Chemicals TRCV92001010G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Vanadium Chloride
CAS 7718-98-1
Molecular Formula VCl3
Quantity 10 g
Synonym Vanadium Trichloride, Vanadium(3+) Chloride, Vanadium(III) Chloride
InChI Formula InChI=1S/3ClH.V/h3*1H;/q;;;+3/p-3
SMILES Cl[V](Cl)Cl
IUPAC Name trichlorovanadium
Molecular Weight (g/mol) 157.3
Formula Weight 155.8505
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.