missing translation for 'onlineSavingsMsg'
Learn More

Vanadium(III) 2,4-pentanedionate, typically 97%

Product Code. 11388217
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11388217 5 g 5g
11378217 25 g 25g
11368217 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11388217 Supplier Thermo Scientific Alfa Aesar Supplier No. 022940.06
 Restricted Item
Estimated Shipment: 09-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 13476-99-8 | C15H21O6V | 348.27 g/mol

Vanadium(III) 2,4-pentanedionate is used as homogeneous catalyst, primary and secondary intermediates, Laboratory chemicals, and manufacture of substances.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13476-99-8
Molecular Formula C15H21O6V
Molecular Weight (g/mol) 348.27
MDL Number MFCD00000033
InChI Key ZVFSRLXFKIWGRT-UHFFFAOYSA-N
Synonym Vanadium(III) acetylacetonate
PubChem CID 122198782
IUPAC Name (Z)-4-hydroxypent-3-en-2-one;vanadium
SMILES [V+3].CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O

Specifications

Quantity 5 g
UN Number UN3285
Sensitivity Air Sensitive
Solubility Information Soluble in water 2g/L,toluene,ethanol,acetone.
Formula Weight 348.27
Percent Purity 97% (typically)
Physical Form Powder
Chemical Name or Material Vanadium(III) 2,4-pentanedionate
compliance-icons
Product Identifier
  • Vanadium(III) 2,4-pentanedionate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 3
Hazard Statement
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H330-Fatal if inhaled.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: Vanadium(III) 2,4-pentanedionate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.