Learn More
Vanadium(III) oxide, 99.7% (metals basis)
Description
Vanadium(III) oxide catalyzes the direct fluorination reactions. It is used in the coating of carbon nanotubes and as an electrode in lithium ion batteries.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 1970°C |
| Physical Form | Powder |
| Quantity | 500 g |
| Assay Percent Range | 99.7% (metals basis) |
| Solubility Information | Slightly soluble in water. |
| Formula Weight | 149.88 |
| Density | 4.87 |
| Chemical Name or Material | Vanadium(III) oxide |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.