missing translation for 'onlineSavingsMsg'
Learn More

Vanadyl phthalocyanine, 85%, pract.

Product Code. 11985871 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11985871 100 g 100g
10568712 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11985871 Supplier Thermo Scientific Acros Supplier No. 422491000
 Restricted Item
Estimated Shipment: 19-02-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 13930-88-6 | C32H16N8OV | 579.46 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13930-88-6
Molecular Formula C32H16N8OV
Molecular Weight (g/mol) 579.46
InChI Key YRZZLAGRKZIJJI-UHFFFAOYSA-N
Synonym oxyvanadium phthalocyanine, vanadium iv oxide phthalocyanine, vanadyl phthalocyanine, vopc, vanadyl iv phthalocyanine, vanadyl phthalocyanine, dye content >90 %
PubChem CID 2735160
SMILES C1=CC=C2C(=C1)C3=NC4=NC(=NC5=C6C=CC=CC6=C([N-]5)N=C7C8=CC=CC=C8C(=N7)N=C2[N-]3)C9=CC=CC=C94.O=[V+2]

Specifications

Color White
Infrared Spectrum Authentic
Assay Percent Range 80 % min
Packaging Glass Bottle
Quantity 100 g
Solubility Information Solubility in water: .. Other solubilities: soluble in xylene,2-ethylhexanoic acid,,2-ethylhexylamine and 6-methyl-2,4-heptanedione
Formula Weight 579.46
Percent Purity 85%
Physical Form Crystalline Powder, Crystals or Chunks
Chemical Name or Material Vanadyl phthalocyanine
compliance-icons
Product Identifier
  • Vanadyl phthalocyanine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.