missing translation for 'onlineSavingsMsg'
Learn More

Vanadyl Sulfate Hydrate, Extra Pure, SLR, Fisher Chemical

Product Code. 10142450 Shop All Fisher Chemical Products
Click to view available options
:
100g, Plastic powder jar
This item is not returnable. View return policy

Product Code. 10142450

Brand: Fisher Chemical V/0105/48

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 12439-96-2
Color Blue
Physical Form Solid
Quantity 100 g
Molecular Formula H10O10SV
MDL Number 149933
InChI Key YGYBXHQARYQUAY-UHFFFAOYSA-L
SMILES O.O.O.O.O.O=[V++].[O-]S([O-])(=O)=O
IUPAC Name oxovanadiumbis(ylium) pentahydrate sulfate
Molecular Weight (g/mol) 253.07
Formula Weight 163g/mol
Packaging Plastic powder jar
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Vanadyl sulfate hydrate
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 1 C
  • Specific target organ toxicity Category 1
Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H361-Suspected of damaging fertility or the unborn child.
  • H372-Causes damage to organs.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contains : Vanadyl sulfate pentahydrate, Vanadyl sulfate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.