missing translation for 'onlineSavingsMsg'
Learn More

Vanillyl Methyl Ketone, TRC

Product Code. 30455068
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30455068 5 g 5g
30450234 1 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30455068 Supplier Toronto Research Chemicals Supplier No. TRCV0976255G
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2503-46-0
Molecular Formula C10H12O3
Molecular Weight (g/mol) 180.2
Synonym 1-(4-Hydroxy-3-methoxyphenyl)-2-propanone, (4-Hydroxy-3-methoxyphenyl)acetone, 1-(4-Hydroxy-3-methoxyphenyl)-2-propanone, 2-Propiovanillone, Guaiacylacetone, Methyl Vanillyl Ketone, NSC 16690
IUPAC Name 1-(4-hydroxy-3-methoxyphenyl)propan-2-one
SMILES COc1cc(CC(=O)C)ccc1O

Specifications

Quantity 5 g
InChI Formula InChI=1S/C10H12O3/c1-7(11)5-8-3-4-9(12)10(6-8)13-2/h3-4,6,12H,5H2,1-2H3
Formula Weight 180.0786
Chemical Name or Material Vanillyl Methyl Ketone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.