missing translation for 'onlineSavingsMsg'
Learn More

Vanillylacetone, TRC

Product Code. 30136855
Change view
Click to view available options
Quantity:
25 g
50 g
100 g
Unit Size:
100g
25g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30136855 50 g 50g
30136704 100 g 100g
30009208 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136855 Supplier Toronto Research Chemicals Supplier No. TRCV09762050G
 Restricted Item
Estimated Shipment: 10-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 122-48-5
Molecular Formula C11 H14 O3
Molecular Weight (g/mol) 194.23
Synonym 4-(4-Hydroxy-3-methoxyphenyl)-2-butanone, 4-Hydroxy-3-methoxyphenyl)ethyl Methyl Ketone, 3-Methoxy-4-hydroxybenzylacetone, 4-(4-Hydroxy-3-methoxyphenyl)-2-butanone, 4-Hydroxy-3-methoxybenzylacetone, Gingerone, NSC 15335, Vanillylacetone, Zingerone, Zingiberone, [0]-Paradol
IUPAC Name 4-(4-hydroxy-3-methoxyphenyl)butan-2-one
SMILES COc1cc(CCC(=O)C)ccc1O

Specifications

Quantity 50 g
InChI Formula InChI=1S/C11H14O3/c1-8(12)3-4-9-5-6-10(13)11(7-9)14-2/h5-7,13H,3-4H2,1-2H3
Formula Weight 194.0943
Chemical Name or Material Zingerone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.