missing translation for 'onlineSavingsMsg'
Learn More

Cole-Parmer™ VapLock™ Solvent Delivery Caps

Product Code. 15854474
Change view
Click to view available options
Size:
GL25
GL45
Material:
PTFE, Polypropylene
PTFE, Polypropylene, Stainless Steel
Number of Channels:
Three Ports
Two Ports
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Material Number of Channels Unit Size
15854474 GL25 PTFE, Polypropylene Two Ports Each
15814524 GL45 PTFE, Polypropylene Three Ports Each
15824524 GL45 PTFE, Polypropylene, Stainless Steel Three Ports Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15854474 Supplier Cole-Parmer™ Supplier No. 1201854
  • ONLINE EXCLUSIVE PRICE
    96.00 €
    65.57 € / Pack of 1
    Save 30.43 €
    31% Off
Estimated Shipment: 17-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

VapLock™ Solvent Delivery Caps reduce chemical evaporation and exposure to hazardous vapors

Use with semi-rigid tubing equal to or less than 1/8 in (3.2 mm) OD

Warranty

None

Specifications

For Use With (Equipment) Solvent-Compatible Containers
Material PTFE, Polypropylene
Number of Channels Two Ports
Size GL25
Thread Size 1/4 in.-28 UNF(F)
For Use With (Application) Solvent Safety
Color Blue/White
Product Type Delivery Cap
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.