Learn More
Cole-Parmer™ VapLock™ Solvent Waste Kits for Bottles and Carboys
Product Code. 15884534
Change view
Click to view available options
Size:
51 mm
GL38
Nalgene 38-430
Wako 38 mm
Material:
PTFE
Polypropylene
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Material unitSize
15884534 GL38 Polypropylene Each
15864534 Nalgene 38-430 Polypropylene Each
15874534 Nalgene 38-430 PTFE Each
15894534 Wako 38 mm Polypropylene Each
17286064 51 mm PTFE Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 15884534 Supplier Cole-Parmer™ Supplier No. WAK38P10
In Stock
Add to basket
Add to basket
VapLock™ Solvent Waste Kits for Bottles and Carboys collect effluent from multiple sources while minimizing harmful solvent exhaust
Various cap sizes available to fit your solvent container
Warranty
None
Specifications
For Use With (Equipment) Solvent-Compatible Containers
Material Polypropylene
Size GL38
Thread Size (English) 1 x 1/2 in. NPT(F), 3 x 1/4 in. NPT(F), 6 x 1/4 in.-28 UN(F)(F)
For Use With (Application) Solvent Safety
Type Solvent Waste Kit