Learn More
Cole-Parmer™ VapLock™ Threaded Plugs
Description
Seal unused manifold and bottle cap ports
Warranty
None
Specifications
Specifications
| For Use With (Equipment) | VapLock Solvent Containment Systems |
| Material | PTFE |
| Thread Size (English) | 1/4 in.-28 UNF(M) |
| For Use With (Application) | Solvent Safety |
| Type | Port Plug |