missing translation for 'onlineSavingsMsg'
Learn More

Varenicline, MedChemExpress

Product Code. 16464246
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16464246 100 mg 100mg
16445577 5 mg 5mg
16432536 25 mg 25mg
16409496 50 mg 50mg
16402838 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16464246 Supplier Medchem Express Supplier No. HY10019
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Varenicline (CP 526555) is a potent partial agonist for α4β2 nicotinic acetylcholine receptor (nAChR) with an EC50 value of 2.3 μM. Varenicline is a full agonist for α3β4 and α7 nAChRs with EC50 values of 55 μM and 18 μM, respectively. Varenicline is a nicotinic ligand based on the structure of cytosine, and has the potential for smoking cessation treatment.

Varenicline (CP 526555) is a potent partial agonist for α4β2 nicotinic acetylcholine receptor (nAChR) with an EC50 value of 2.3 μM. Varenicline is a full agonist for α3β4 and α7 nAChRs with EC50 values of 55 μM and 18 μM, respectively. Varenicline is a nicotinic ligand based on the structure of cytosine, and has the potential for smoking cessation treatment.

Specifications

Chemical Name or Material Varenicline
CAS 249296-44-4
Color Yellow
Purity Grade Notes Research
Molecular Formula C13H13N3
Quantity 100 mg
Synonym CP 526555
Solubility Information DMSO : 25 mg/mL (118.34 mM; Need ultrasonic)
SMILES C12=C(C=C3N=CC=NC3=C2)C4CC1CNC4
Molecular Weight (g/mol) 211.26
Formula Weight 211.26
Percent Purity 99.98%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.