Learn More
Biochrom Variable Pathlength Cell Holder
Product Code. 11603024 Shop All Harvard Bioscience Products
Change view
Click to view available options
Type:
Cell Holder
Unit Size:
Each
Product Code. Type Unit Size
11603024 Cell Holder Each
1 options
Product Code. 11603024 Supplier Biochrom Supplier No. 80710005
  • ONLINE EXCLUSIVE PRICE
    1110.00 €
    1031.52 € / Pack of 1
    Save 78.48 €
    7% Off
Estimated Shipment: 06-01-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Variable Pathlength Cell Holder for use with Ultrospec 7000/8000/9000.
  • Holds 1, 2, 5, 10, 20, 50 and 100 mm pathlength cells
  • Includes one thumb screw
Specifications
Type Cell Holder
Disposable Reusable
For Use With (Equipment) Ultrospec 7000/8000/9000
Holds 1, 2, 5, 10, 20, 50 and 100 mm Pathlength Cells
Includes 1 x Thumb Screw
Shape Rectangular