Learn More
Hamilton™ Bruker Varian™ Replacement Needles
Product Code. 10287711
Product Code. 10287711
Brand: Hamilton™ 202903
For use with Bruker Varian Autosamplers.
  • 10μL volume
  • 26s gauge
  • 50.5mm in length
  • Point style 5