missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Varioskan™ LUX multimode microplate reader, Absorbance, Fluorescence intensity, Luminescence and AlphaScreen

Product Code. 15300777 Shop All Thermo Scientific Products
Change view
Click to view available options
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. unitSize
15300777 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15300777 Supplier Thermo Scientific™ Supplier No. VL0LA0D2

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Analyze a variety of samples with five measurement technologies, including absorbance (UV-Vis), fluorescence intensity, Luminescence, AlphaScreen / AlphaLISA and TRF.

Designed for bioscience researchers with a wide variety of needs, the Thermo Scientific™ Varioskan™ LUX multimode microplate reader comes equipped with a flexible range of measurement technologies including Absorbance, Fluorescence Intensity, Luminescence, AlphaScreen, and Time-Resolved Fluorescence. The instrument simplifies measurement setup with the automatic dynamic range selection, and its smart safety controls ease workflow and help avoid experimental errors. Varioskan LUX raises the bar for reliability and ease, with technology that works in researchers' best interest.

The Varioskan LUX multimode reader has received HTRF™ certification from Cisbio Bioassays and DLReady™ validation from Promega Corporation.

The Varioskan LUX is a modular, upgradable instrument that provides excellent usability across a variety of applications.

  • Up to five measurement technologies: Absorbance (UV-Vis), Fluorescence intensity (incl. FRET), Luminescence (direct and filtered), AlphaScreen/AlphaLISA and Time-resolved fluorescence (incl. TR-FRET/hTRF)
  • Filter and monochromator optics
  • Spectral scanning for assay optimization
  • Five measurement modes including end point, kinetic, spectra, kinetic spectra and multipoint
  • Integrated gas control module for setting CO2 and O2 concentrations for cell based assays
  • Simultaneous dispensing and measurement for signal monitoring at the start of a reaction
  • Automatic dynamic range selection, based on signal intensity of each well
  • Smart safety controls designed to identify and alert the user of potential problems before they happen
  • Automated instrument calibration and self-diagnostics for reliable performance
  • Certified HTRF compatible reader by CisBio Bioassays
  • Validated DLReady by Promega Corporation

Varioskan LUX is paired with the powerful and intuitive SkanIt Software, providing excellent usability and flexibility.

  • License free—Each user can install the software on his/her computer
  • Intuitive and flexible interface for multiple users and assays
  • Innovative Virtual Pipetting Tool makes it easy to define samples to plate layout
  • Measurement data is continuously saved to the database, so there's no data lost due to unexpected interruptions such as power outages or accidental aborting
  • Simplified data processing and export
    • Export data in a single mouse click, or have it emailed automatically at the end of the run
    • Several file formats for exporting results: *.xlsx, *.pdf, *.xml, and *.txt
  • Shows real-time spectra and kinetic curves during the measurement
  • Built-in calculations for data analysis (e.g. blank subtraction, average/SD/CV%, standard curve, dose response, quality control, classification, kinetic and spectral analysis, custom formula, etc.)
  • Comprehensive result reports with traceability to runtime actions and settings
  • Two editions available: Research Edition for life science researchers, and Drug Discovery Edition which offers features needed for the FDA 21 CFR Part 11 compliance.

The Varioskan LUX is modular and upgradeable, so you can build the system that best suits your needs. For more information, contact a sales representative.

TRUSTED_SUSTAINABILITY

Specifications

Compatibility Robot
Detector Type Photomultiplier Tube (PMT)
Incubator Temperature 45°C
Light Source Xenon Flash Lamp, LED
Measurement Speed 96 wells: 15 sec., 384 wells: 45 sec., 1536 wells: 135 sec.
Measurement Technology Absorbance, Fluorescence Intensity, Luminescence, AlphaScreen
Plate Type 6 to 1536-well Plates (Absorbance 6 to 384-well Plates)
Product Line Varioskan
Reading Options Top
Read-out Range 0 to 6 Abs
Shaking Orbital
Software Type SkanIt Software
Spectral Scanning Speed < 2.2 sec./well 400 to 500 nm, 1 flash, 2 nm Step
Wavelength Selection Double monochromaters, Double excitation monochromaters, Double emission monochromators, Filters
Accuracy 0.003 Abs or ±2%, at 200 to 399 nm (0 to 2 Abs)
0.003 Abs or ±1%, at 400 to 1000 nm (0 to 3 Abs)
Depth (English) 23 in.
Depth (Metric) 58 cm
Dispense Volume 1 mL syringe: 2 to 5,000 μL
5 mL syringe: 5 to 25,000 μL
Dynamic Range Fluorescence intensity: top reading > 6 decades
Luminescence: > 7 decades
Height (English) 20 in.
Height (Metric) 51 cm
Linearity 96-well plate: ±2% (0 to 4 Abs at 450 nm)
384-well plate: ±2% (0 to 3 Abs at 450 nm)
Measurement Type Endpoint, Kinetic, Spectra, Multipoint, Kinetic Spectra
No. of Dispensers 2
Precision SD < 0.001 Abs or CV < 0.5%, at 450 nm (0 to 3 Abs)
Sensitivity Fluorescence intensity (top reading): < 0.4 fmol fluorescein/well (black 384-well plate)
Luminescence: < 7 amol ATP/well (white 384-well plate)
AlphaScreen: < 100 amol phosphotyrosine/well (384-well plate)
Software Language Versions English, German, French, Spanish, Portuguese, Italian, Japansese, Chinese and Russian
Wavelength Range Absorbance: 200 to 1000 nm
Fluorescence intensity: Excitation: 200 to 1000 nm, Emission: 270 to 840 nm
Luminescence: 360 to 670 nm
AlphaScreen: Excitation 680 nm, Emission 400 to 660 nm
Weight (English) 130 lb.
Weight (Metric) 59 kg
Width (English) 21 in.
Width (Metric) 53 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.