missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Varioskan™ LUX Multimode Microplate Reader

Product Code. 17153461 Shop All Thermo Scientific Products
Click to view available options
Measurement Technology:
Absorbance, Fluorescence Intensity
Absorbance, Fluorescence Intensity, Luminescence
Absorbance, Fluorescence Intensity, Luminescence, Time-resolved Fluorescence
Absorbance, Fluorescence Intensity, Luminescence, Time-resolved Fluorescence, AlphaScreen
Reading Options:
Top
Top and Bottom
No. of Dispensers:
1
2
None
Includes:
Gas Module, SkanIt PC Software, Research Edition
SkanIt PC Software, Research Edition
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 17153461

Brand: Thermo Scientific™ VLBLATGD2

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Analyze a variety of samples with five measurement technologies, including absorbance (UV-Vis), fluorescence intensity, Luminescence, AlphaScreen / AlphaLISA and TRF.

Designed for bioscience researchers with a wide variety of needs, the Thermo Scientific™ Varioskan™ LUX multimode microplate reader comes equipped with a flexible range of measurement technologies including Absorbance, Fluorescence Intensity, Luminescence, AlphaScreen, and Time-Resolved Fluorescence. The instrument simplifies measurement setup with the automatic dynamic range selection, and its smart safety controls ease workflow and help avoid experimental errors. Varioskan LUX raises the bar for reliability and ease, with technology that works in researchers' best interest.

The Varioskan LUX multimode reader has received HTRF™ certification from Cisbio Bioassays and DLReady™ validation from Promega Corporation.

The Varioskan LUX is a modular, upgradable instrument that provides excellent usability across a variety of applications.

  • Up to five measurement technologies: Absorbance (UV-Vis), Fluorescence intensity (incl. FRET), Luminescence (direct and filtered), AlphaScreen/AlphaLISA and Time-resolved fluorescence (incl. TR-FRET/hTRF)
  • Filter and monochromator optics
  • Spectral scanning for assay optimization
  • Five measurement modes including end point, kinetic, spectra, kinetic spectra and multipoint
  • Integrated gas control module for setting CO2 and O2 concentrations for cell based assays
  • Simultaneous dispensing and measurement for signal monitoring at the start of a reaction
  • Automatic dynamic range selection, based on signal intensity of each well
  • Smart safety controls designed to identify and alert the user of potential problems before they happen
  • Automated instrument calibration and self-diagnostics for reliable performance
  • Certified HTRF compatible reader by CisBio Bioassays
  • Validated DLReady by Promega Corporation

Varioskan LUX is paired with the powerful and intuitive SkanIt Software, providing excellent usability and flexibility.

  • License free—Each user can install the software on his/her computer
  • Intuitive and flexible interface for multiple users and assays
  • Innovative Virtual Pipetting Tool makes it easy to define samples to plate layout
  • Measurement data is continuously saved to the database, so there's no data lost due to unexpected interruptions such as power outages or accidental aborting
  • Simplified data processing and export
    • Export data in a single mouse click, or have it emailed automatically at the end of the run
    • Several file formats for exporting results: *.xlsx, *.pdf, *.xml, and *.txt
  • Shows real-time spectra and kinetic curves during the measurement
  • Built-in calculations for data analysis (e.g. blank subtraction, average/SD/CV%, standard curve, dose response, quality control, classification, kinetic and spectral analysis, custom formula, etc.)
  • Comprehensive result reports with traceability to runtime actions and settings
  • Two editions available: Research Edition for life science researchers, and Drug Discovery Edition which offers features needed for the FDA 21 CFR Part 11 compliance.

The Varioskan LUX is modular and upgradeable, so you can build the system that best suits your needs. For more information, contact a sales representative.

TRUSTED_SUSTAINABILITY

Specifications

Compatibility Robot
Detector Type Photomultiplier Tube (PMT)
Dimensions (L x W x H) 58 x 53 x 51 cm (23 x 21 x 20 in.)
Incubator Temperature Ambient +4° to 45°C
Interface PC Software (Thermo Scientific SkanIt Software)
Light Source Xenon Flash Lamp, LED
Measurement Speed 15 s for 96-well Plate, 45 s for 384-well Plate, 135 s for 1536-Well Plate (Minimum Times)
Measurement Technology Absorbance, Fluorescence Intensity, Luminescence, Time-resolved Fluorescence, AlphaScreen
Plate Type 6 to 1536-well Plates (Absorbance 6 to 384-well Plates), μDrop Plates
Product Line Varioskan
Reading Options Top and Bottom
Read-out Range 0 to 6 Abs
Shaking Orbital
Software Type SkanIt Software
Spectral Scanning Speed < 2.2 s/Well 400 to 500 nm, 1 Flash, 2 nm Step
Wavelength Selection Double Monochromator (Absorbance and Spectral Scanning), Double Excitation and Emission Monochromator (Fluorescence Intensity), Filter (AlphaScreen and Time-Resolved Fluorescence)
Accuracy (Photometric) 0.003 Abs or ±2% at 200 to 399 nm (0 to 2 Abs), 0.003 Abs or ±1%, at 400 to 1000 nm (0 to 3 Abs)
CO2 Concentration Range 0.1 to 15%
Depth (English) 23 in.
Depth (Metric) 58 cm
Dispense Volume 2 – 5 000 μl with 1 μl increments
Dynamic Range Fluorescence intensity: top reading > 6 decades; Luminescence: > 7 decades; Time-resolved fluorescence: > 6 decade
Emission Filters Available for purchase
Excitation Filters Fixed filters (Alpha Screen/AlphaLISA excitation wavelength 680 nm; TRF 334 nm)
Height (English) 20 in.
Height (Metric) 51 cm
Includes Gas Module, SkanIt PC Software, Research Edition
Linearity 96-well plate: 0 to 4 Abs, ±2% at 450 nm, 384-well plate: 0 to 3 Abs ±2% at 450 nm
Measurement Type Endpoint, Kinetic, Spectra, Multipoint, Kinetic Spectra
No. of Dispensers 2
O2 Concentration Range 1 to 21%
Precision SD <0.001 Abs or CV <0.5%, at 450 nm (0 to 3 Abs)
Sensitivity Fluorescence intensity (top reading): < 0.4 fmol fluorescein/well (black 384-well plate), Fluorescence intensity (bottom reading): < 4 fmol fluorescein/well, (clear bottom black 384 square well plate), Time-resolved fluorescence: < 1 amol/well (white low volume 384 well plate)Luminescence: < 7 amol ATP/well (white 384-well plate), AlphaScreen: < 100 amol phosphotyrosine/well (384-well plate)
Software Language Versions English, German, French, Spanish, Portuguese, Italian, Japansese, Chinese
Wavelength Range Absorbance: 200-1000 nm; Fluorescence intensity: Excitation: 200-822 nm, Emission: 270-840 nm: TRF: Excitation 334 nm, Emission 400 to 700 nm; Luminescence: 360-670 nm; AlphaScreen: Excitation 680 nm, Emission 400-660 nm
Weight (English) 125 lb
Weight (Metric) 57 kg
Width (English) 21 in.
Width (Metric) 53 cm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.