missing translation for 'onlineSavingsMsg'
Learn More

Vatalanib Dihydrochloride, TRC

Product Code. 30456988
Change view
Click to view available options
Quantity:
10 mg
25 mg
Unit Size:
10mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30456988 10 mg 10mg
30450552 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30456988 Supplier Toronto Research Chemicals Supplier No. TRCV09950010MG
 Restricted Item
Estimated Shipment: 17-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 212141-51-0
Molecular Formula C20 H15 Cl N4 . 2 Cl H
Molecular Weight (g/mol) 419.73
Synonym 1-Phthalazinamine, N-(4-chlorophenyl)-4-(4-pyridinylmethyl)-, hydrochloride (1:2), 1-Phthalazinamine, N-(4-chlorophenyl)-4-(4-pyridinylmethyl)-, dihydrochloride (9CI), Vatalanib dihydrochloride
IUPAC Name N-(4-chlorophenyl)-4-(pyridin-4-ylmethyl)phthalazin-1-amine;dihydrochloride
SMILES Cl.Cl.Clc1ccc(Nc2nnc(Cc3ccncc3)c4ccccc24)cc1

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C20H15ClN4.2ClH/c21-15-5-7-16(8-6-15)23-20-18-4-2-1-3-17(18)19(24-25-20)13-14-9-11-22-12-10-14;;/h1-12H,13H2,(H,23,25);2*1H
Formula Weight 418.0519
Chemical Name or Material Vatalanib Dihydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.