Learn More
Sonics Materials VC 750 Ultrasonic Processor
Product Code. 11743619
Change view
Click to view available options
Wattage:
750 W
Unit Size:
Each
Product Code. Wattage Unit Size
11743619 750 W Each
1 options
Product Code. 11743619 Supplier Sonics Materials Supplier No. VC750220
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
missing translation for 'validMainframeMsg'
Robust and reliable 750 Watt ultrasonic processors for small and medium volume applications that are ideal for safely processing a wide range of organic and inorganic materials from 250 microliters to 1 liter.
  • Ultrasonic processors for small and medium volume applications
  • 750 watt ultrasonic processors to safely process a wide range of organic and inorganic materials, from 250 microliters to 1 liter
  • Typical applications include nanotechnology (producing nanoparticle materials and Graphene dispersions), cell lysing, sample preparation, homogenization, ChIP Assay, emulsification, disaggregation and deagglomeration, plus uses in the field of sonochemical liquid processing
  • Energy monitor, digital wattmeter, microprocessor-based and programmable
  • 10 hour process timer
Specifications
Product Type Ultrasonic Processor
Wattage 750 W