Learn More
Sonics Materials VCX-130PB Ultrasonic Processor
Product Code. 11763729
Change view
Click to view available options
Wattage:
130 W
Unit Size:
Each
Product Code. Wattage Unit Size
11763729 130 W Each
1 options
Product Code. 11763729 Supplier Sonics Materials Supplier No. VCX130PB220
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
missing translation for 'validMainframeMsg'
Robust and reliable 130 watt Ultrasonic Processor with thumb-actuated pulser for small volume applications.
  • Ultrasonic processors for small volume applications
  • 130 watt ultrasonic processor with thumb-actuated pulser or footswitch
  • Safely process a wide range of organic and inorganic materials, from 150 microliters to 150 milliliters
  • Typical applications include biotechnology and pharmaceutical processing, including mixing, dispersion, and sample prep uses
  • Energy monitor, digital wattmeter, automatic tuning
  • Variable power output control
Specifications
Product Type Ultrasonic Processor
Wattage 130 W
Includes Thumb-actuated pulser