Learn More
Sonics Materials VCX-1500HV Ultrasonic Processor
Product Code. 11783649
Change view
Click to view available options
Wattage:
1500 W
Unit Size:
Each
Product Code. Wattage unitSize
11783649 1500 W Each
1 options
Product Code. 11783649 Supplier Sonics Materials Supplier No. VCX1500HV220
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Designed primarily for dispersing and homogenizing, this 1500 Watt high volume ultrasonic liquid processing system is ideal for large volume applications.
  • Ultrasonic processors and equipment for large volume applications
  • The flow cell throughput rate is typically 100 liters/hour – variables being viscosity and desired degree of processing
  • The flow cell is recommended for the treatment of low viscosity samples which do not require extended exposure to ultrasonics
  • Designed primarily for dispersing and homogenizing
  • The flow cell is easily disassembled for inspection and cleaning, and is water jacketed to enable the sample to be cooled/heated while it is being processed
Spécification
Wattage 1500 W
Nom du produit
Sélectionnez un problème
En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.