missing translation for 'onlineSavingsMsg'
Learn More

Vegita, TRC

Product Code. 30012519
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30012519 1 mg 1mg
30004927 10 mg 10mg
30009037 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30012519 Supplier Toronto Research Chemicals Supplier No. TRCV1100001MG
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 33813-20-6
Molecular Formula C4 H4 N2 S3
Molecular Weight (g/mol) 176.28
Synonym 3H-Imidazo[2,1-c]-1,2,4-dithiazole-3-thione, 5,6-dihydro-, 5,6-Dihydro-3H-imidazo[2,1-c]-1,2,4-dithiazole-3-thione, EBIS, ETM, ETM (heterocycle), Endodan, Etem, Ethylenebisdiisothiocyanato sulfide, Ethylenebisthiuram monosulfide, Ethylenethiocarbamyl sulfide, Ethylenethiuram monosulfide, Ethylenethiuram sulfide, Hortocritt, N,N'-Dimethylene-5-imino-1,2-dithio-4-azolidine-3-thione, Thiuram monosulfide, 1,2-ethanediyl-, Vegita
IUPAC Name 5,6-dihydroimidazo[2,1-c][1,2,4]dithiazole-3-thione
SMILES S=C1SSC2=NCCN12

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C4H4N2S3/c7-4-6-2-1-5-3(6)8-9-4/h1-2H2
Formula Weight 175.9537
Chemical Name or Material EBIS
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.