missing translation for 'onlineSavingsMsg'
Learn More

Velsecorat, MedChemExpress

Product Code. 16497177
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16497177 5 mg 5mg
16495336 100 mg 100mg
16454458 10 mg 10mg
16433096 25 mg 25mg
16400777 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16497177 Supplier Medchem Express Supplier No. HY111453
 Restricted Item
Estimated Shipment: 19-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress AZD7594 is a potent selective nonsteroidal glucocorticoid receptor modulator, with an IC50 of 0.9 nM.

AZD7594 is a potent selective nonsteroidal glucocorticoid receptor modulator, with an IC50 of 0.9 nM.

Specifications

Chemical Name or Material Velsecorat
CAS 1196509-60-0
Color White
Purity Grade Notes Research
Molecular Formula C32H32F2N4O6
Quantity 5 mg
Synonym AZD7594 AZ13189620
Solubility Information DMSO : ≥ 83.3 mg/mL (137.32 mM)
SMILES O=C(N[C@H]1COCC1)C2=CC=CC(N3N=CC4=C3C=CC(O[C@H](C5=CC=C(OCCO6)C6=C5)[C@@H](NC(C(F)(F)C)=O)C)=C4)=C2
Molecular Weight (g/mol) 606.62
Formula Weight 606.62
Percent Purity 99.6%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.