missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Vemurafenib

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
Vemurafenib is a potent inhibitor of BRAFV600E, C-Raf and wild type B-Raf (IC50 values are 13-31 nM, 6.7-48 nM and 100-160 nM, respectively. Vemurafenib also inhibits SRMS, ACK1 KHS1, FGR (IC50 values are 18 nM, 19 nM, 51 nM and 63 nM, respectively). Vemurafenib shows antitumor activity and induces autophagy in BRAF-mutant thyroid cancer cells.
Specifications
Specifications
| Chemical Name or Material | N-[3-[[5-(4-Chlorophenyl)-1 H-pyrrolo[2,3-b]pyridin-3-yl]carbonyl]-2,4-difluorophenyl]-1-propanesulfonamide |
| CAS | 918504-65-1 |
| Quantity | 10 mg |
| Target | Raf Kinase Inhibitors |
| Molecular Formula | C23H18ClF2N3O3S |
| Purity | 0.98 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction